Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 3.10 mm, 2.000 in
Catalog No. 5000527894
Encompass
This plug gage is designed for precise measurements and calibration tasks. Made from tool steel, it features a 3.10 mm diameter and an overall length of 2.000 in. It is finished to 10 micro and complies with ASME B89.1 standards, making it suitable for a variety of inspection and calibration applications.
  • For go inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-27894 Supplier MSC Supplier No. 68094150
This plug gage is designed for precise measurements and calibration tasks. Made from tool steel, it features a 3.10 mm diameter and an overall length of 2.000 in. It is finished to 10 micro and complies with ASME B89.1 standards, making it suitable for a variety of inspection and calibration applications.
  • For go inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability