This plug gage is a Class ZZ Plus model, featuring a 3.27 mm diameter and an overall length of 2.0000 inches. It is constructed from durable tool steel with a 10 Micro finish and offers a tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST Traceability, making it ideal for "Go" inspection and for precisely setting micrometers, verniers, and other measuring devices in accordance with ASME B89.1 standards.
- Class ZZ Plus for precise measurements.
- 3.27 mm diameter.
- 2.0000 inches (50.8 mm) overall length.
- Tolerance of 0.00508 mm.
- Tool steel construction.
- 10 Micro finish.
- Includes certificate of accuracy with NIST traceability.
- Suitable for "Go" inspection.
- Used for setting micrometers, verniers, and other measuring devices.
- Complies with ASME B89.1 standards.