Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 3.32 mm Diameter, Tool Steel
Catalog No. 5000530664
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and calibration in laboratory and industrial settings. Constructed from durable tool steel, it offers accurate verification for a range of instruments.
  • For go inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Diameter of measurement 3.32 mm
  • Overall length 2.0000 in
  • Tolerance 0.00508 mm
  • Finish 10 micro
Catalog No. 50-005-30664 Supplier MSC Supplier No. 68094374
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and calibration in laboratory and industrial settings. Constructed from durable tool steel, it offers accurate verification for a range of instruments.
  • For go inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Diameter of measurement 3.32 mm
  • Overall length 2.0000 in
  • Tolerance 0.00508 mm
  • Finish 10 micro