Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 3.57 mm Diameter, Tool Steel
Catalog No. 5000527909
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in various applications. It features a 3.57 mm diameter and is constructed from durable tool steel, ensuring accuracy and longevity. Ideal for go inspection and setting a range of measuring devices, it comes with a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • For go inspection applications
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Constructed from durable tool steel
  • Features a precision diameter of 3.57 mm
  • Overall length of 2.0000 in
Catalog No. 50-005-27909 Supplier MSC Supplier No. 68094622
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in various applications. It features a 3.57 mm diameter and is constructed from durable tool steel, ensuring accuracy and longevity. Ideal for go inspection and setting a range of measuring devices, it comes with a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • For go inspection applications
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Constructed from durable tool steel
  • Features a precision diameter of 3.57 mm
  • Overall length of 2.0000 in