Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 4.10 mm Diameter, Tool Steel
Catalog No. 5000530953
Encompass
This precision plug gage is manufactured by Vermont Gage, ideal for accurate 'Go' inspection and calibrating various measuring instruments. Crafted from durable tool steel, it ensures consistent performance and reliability in demanding applications.
  • Features Class ZZ Plus tolerance for exceptional accuracy.
  • Constructed from high-quality tool steel for enhanced durability.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Conforms to ASME B89.1 standards for quality assurance.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Boasts a 10 micro finish for smooth operation.
Catalog No. 50-005-30953 Supplier MSC Supplier No. 68095157
This precision plug gage is manufactured by Vermont Gage, ideal for accurate 'Go' inspection and calibrating various measuring instruments. Crafted from durable tool steel, it ensures consistent performance and reliability in demanding applications.
  • Features Class ZZ Plus tolerance for exceptional accuracy.
  • Constructed from high-quality tool steel for enhanced durability.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Conforms to ASME B89.1 standards for quality assurance.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Boasts a 10 micro finish for smooth operation.