Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 4.14 mm Diameter, 2.0000 in OAL
Catalog No. 5000531388
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in demanding applications. Featuring a 4.14 mm diameter and a 2.0000 inch overall length, it is constructed from durable tool steel. This gage is ideal for 'Go' inspection and for setting micrometers, verniers, and other measuring devices, ensuring accuracy and reliability.
  • Has a 0.00508 mm tolerance
  • Features a 10 micro finish
  • Designed for Go inspection; setting micrometers; verniers & other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel
Catalog No. 50-005-31388 Supplier MSC Supplier No. 68095199
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in demanding applications. Featuring a 4.14 mm diameter and a 2.0000 inch overall length, it is constructed from durable tool steel. This gage is ideal for 'Go' inspection and for setting micrometers, verniers, and other measuring devices, ensuring accuracy and reliability.
  • Has a 0.00508 mm tolerance
  • Features a 10 micro finish
  • Designed for Go inspection; setting micrometers; verniers & other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Constructed from tool steel