Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 4.45 mm Dia, 2.0000 in OAL
Catalog No. 5000536328
Encompass
This plug gage is engineered for precise measurement tasks, featuring a 4.45 mm diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is ideal for 'Go' inspection and calibrating micrometers, verniers, and other measuring instruments.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for 'Go' inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Features a tolerance of 0.00508 mm.
  • Has a 10 micro finish.
  • Manufactured from tool steel.
  • Complies with ASME B89.1 standards.
Catalog No. 50-005-36328 Supplier MSC Supplier No. 68095504
This plug gage is engineered for precise measurement tasks, featuring a 4.45 mm diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is ideal for 'Go' inspection and calibrating micrometers, verniers, and other measuring instruments.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for 'Go' inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Features a tolerance of 0.00508 mm.
  • Has a 10 micro finish.
  • Manufactured from tool steel.
  • Complies with ASME B89.1 standards.