Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.04 mm Diameter, 0.00508 mm Tolerance
Catalog No. 5000527914
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is engineered for precision measurement and inspection. Constructed from durable tool steel, this plug gage features a 5.04 mm diameter, a 0.00508 mm tolerance, and a 10 microfinish. With an overall length of 2.0000 in (50.8 mm), it is ideal for Go inspection and setting various measuring devices like micrometers and verniers, adhering to ASME B89.1 standards.
  • Application for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 microfinish.
  • Constructed from tool steel.
  • Overall length of 2.0000 in (50.8 mm).
Catalog No. 50-005-27914 Supplier MSC Supplier No. 68096098
The Vermont Gage Class ZZ Plus Plug Gage is engineered for precision measurement and inspection. Constructed from durable tool steel, this plug gage features a 5.04 mm diameter, a 0.00508 mm tolerance, and a 10 microfinish. With an overall length of 2.0000 in (50.8 mm), it is ideal for Go inspection and setting various measuring devices like micrometers and verniers, adhering to ASME B89.1 standards.
  • Application for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 microfinish.
  • Constructed from tool steel.
  • Overall length of 2.0000 in (50.8 mm).