Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.27 mm, Tool Steel, ASME B89.1
Catalog No. 5000530954
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a 5.27 mm diameter and meets ASME B89.1 standards. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for critical measurements.
  • For go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Adheres to ASME B89.1 standards
  • Features a 0.00508 mm tolerance
  • Coated with a 10 micro finish
Catalog No. 50-005-30954 Supplier MSC Supplier No. 68096320
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a 5.27 mm diameter and meets ASME B89.1 standards. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for critical measurements.
  • For go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Adheres to ASME B89.1 standards
  • Features a 0.00508 mm tolerance
  • Coated with a 10 micro finish