Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.28 mm Diameter, Tool Steel
Catalog No. 5000527244
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and calibrating measuring devices. Made from durable tool steel, it features a 5.28 mm diameter and an overall length of 2.0000 inches, ensuring high accuracy with a tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Precise tolerance of 0.00508 mm.
  • Coating/finish of 10 micro.
  • Overall length of 2.0000 in.
  • Constructed from durable tool steel.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Measuring diameter is 5.28 mm.
  • Manufactured in the United States.
Catalog No. 50-005-27244 Supplier MSC Supplier No. 68096338
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and calibrating measuring devices. Made from durable tool steel, it features a 5.28 mm diameter and an overall length of 2.0000 inches, ensuring high accuracy with a tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Precise tolerance of 0.00508 mm.
  • Coating/finish of 10 micro.
  • Overall length of 2.0000 in.
  • Constructed from durable tool steel.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Measuring diameter is 5.28 mm.
  • Manufactured in the United States.