Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.37 mm Dia, 2.0000 in OAL
Supplier:  MSC 68096429
Encompass
This Class ZZ Plus Plug Gage features a precision design for accurate measurements. With a 5.37 mm diameter and a 2.0000 inch overall length, it is constructed from durable Tool Steel and boasts a 10 Micro finish. Ideal for Go Inspection, this gage is suitable for setting micrometers, verniers, and other measuring devices, and includes a Certificate of Accuracy with NIST Traceability, ensuring compliance with ASME B89.1 standards.
  • Precision Class ZZ Plus design
  • 5.37 mm diameter for precise measurements
  • 2.0000 inch overall length
  • Constructed from durable tool steel
  • Features a 10 Micro coating/finish
  • Designed for Go Inspection applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Adheres to ASME B89.1 standards
  • Manufactured in the United States
Catalog No. 50-005-27907