Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.75 mm Diameter, 2.000 in OAL
Catalog No. 5000527908
Encompass
This Class ZZ Plus Plug Gage is an essential tool for precise Go Inspection and for accurately setting micrometers, verniers, and other measuring devices. It offers a 5.75 mm diameter and a 2.0000 inch overall length, constructed from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Application for Go Inspection.
  • Sets micrometers, verniers, and other measuring devices.
Catalog No. 50-005-27908 Supplier MSC Supplier No. 68096809
This Class ZZ Plus Plug Gage is an essential tool for precise Go Inspection and for accurately setting micrometers, verniers, and other measuring devices. It offers a 5.75 mm diameter and a 2.0000 inch overall length, constructed from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Application for Go Inspection.
  • Sets micrometers, verniers, and other measuring devices.