Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.76 mm, Tool Steel
Catalog No. 5000527242
Encompass
This plug gage is a Class ZZ Plus gage with a 5.76 mm diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is designed for use in go inspection and for accurately setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from tool steel for durability.
  • Features a 10 micro finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-27242 Supplier MSC Supplier No. 68096817
This plug gage is a Class ZZ Plus gage with a 5.76 mm diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is designed for use in go inspection and for accurately setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from tool steel for durability.
  • Features a 10 micro finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.