Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 5.82 mm, Tool Steel, 10 Microfinish
Catalog No. 5000532005
Encompass
The Class ZZ Plus Plug Gage is a precision instrument for accurate measurement and inspection. With a 5.82 mm diameter and constructed from durable tool steel, it features a 10 microfinish for reliable performance. This gage is essential for go inspection and for setting various measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-32005 Supplier MSC Supplier No. 68096874
The Class ZZ Plus Plug Gage is a precision instrument for accurate measurement and inspection. With a 5.82 mm diameter and constructed from durable tool steel, it features a 10 microfinish for reliable performance. This gage is essential for go inspection and for setting various measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.