Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 7.55 mm, Tool Steel
Catalog No. 5000527330
Encompass
This Class ZZ Plus Plug Gage is crafted from durable tool steel for precise "Go" inspection. It features a 7.55 mm diameter and 2.0000 inch overall length. With a 0.00508 mm tolerance and 10 Micro coating/finish, it's ideal for calibrating micrometers, verniers, and other measuring instruments. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Designed for "Go" inspection
  • Calibrates micrometers, verniers, and other measuring devices
  • Includes Certificate of Accuracy with NIST Traceability
  • Conforms to ASME B89.1 standards
  • Tool steel construction
  • 7.55 mm measurement diameter
  • 2.0000 inch overall length
  • 0.00508 mm tolerance
  • 10 Micro coating/finish
Catalog No. 50-005-27330 Supplier MSC Supplier No. 68098607
This Class ZZ Plus Plug Gage is crafted from durable tool steel for precise "Go" inspection. It features a 7.55 mm diameter and 2.0000 inch overall length. With a 0.00508 mm tolerance and 10 Micro coating/finish, it's ideal for calibrating micrometers, verniers, and other measuring instruments. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Designed for "Go" inspection
  • Calibrates micrometers, verniers, and other measuring devices
  • Includes Certificate of Accuracy with NIST Traceability
  • Conforms to ASME B89.1 standards
  • Tool steel construction
  • 7.55 mm measurement diameter
  • 2.0000 inch overall length
  • 0.00508 mm tolerance
  • 10 Micro coating/finish