Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 8.72 mm Diameter, Tool Steel
Catalog No. 5000527273
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection. Constructed from durable tool steel, this 8.72 mm diameter gage is ideal for go inspection and setting various measuring devices. It adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ Plus for high precision
  • 8.72 mm diameter for accurate measurements
  • Made from durable tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Compliant with ASME B89.1 standards
  • 10 Micro coating/finish for enhanced durability
Catalog No. 50-005-27273 Supplier MSC Supplier No. 68099779
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection. Constructed from durable tool steel, this 8.72 mm diameter gage is ideal for go inspection and setting various measuring devices. It adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ Plus for high precision
  • 8.72 mm diameter for accurate measurements
  • Made from durable tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Compliant with ASME B89.1 standards
  • 10 Micro coating/finish for enhanced durability