Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 9.63 mm Diameter
Catalog No. 5000510660
Encompass
This plug gage is engineered for precise measurement applications. It features a 9.63 mm diameter, a Class ZZ Plus designation, and a strict 0.00508 mm tolerance. With an overall length of 2.000 inches, this gage is suitable for various calibration and inspection tasks requiring high accuracy.
  • 9.63 mm diameter
  • Class ZZ Plus tolerance
  • 2.000 inch overall length
  • 0.00508 mm tolerance
  • 10 micro finish
Catalog No. 50-005-10660 Supplier MSC Supplier No. 68100684
This plug gage is engineered for precise measurement applications. It features a 9.63 mm diameter, a Class ZZ Plus designation, and a strict 0.00508 mm tolerance. With an overall length of 2.000 inches, this gage is suitable for various calibration and inspection tasks requiring high accuracy.
  • 9.63 mm diameter
  • Class ZZ Plus tolerance
  • 2.000 inch overall length
  • 0.00508 mm tolerance
  • 10 micro finish