Learn More
MSC Vermont Gage Plug Gage Set, 5 to 9.98 mm, Class ZZ, 250 Pc
Catalog No. 5000528429
Encompass
This Class ZZ plug gage set by Vermont Gage is designed for precision measurement tasks. It covers a range from 5.00 mm to 9.98 mm, with a tolerance of -0.005 mm, and includes 250 individual pieces.
  • Covers a diameter range from 5.00 mm to 9.98 mm.
  • Features a class ZZ tolerance of 0.0050 mm.
  • Includes 250 individual steel gages.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for NO-GO gaging inspection.
  • Ideal for gaging slot widths and locations, and measuring hole diameters.
  • Comes with a certificate of accuracy with NIST traceable standards.
  • Manufactured in the United States.
Catalog No. 50-005-28429 Supplier MSC Supplier No. 40427338
This Class ZZ plug gage set by Vermont Gage is designed for precision measurement tasks. It covers a range from 5.00 mm to 9.98 mm, with a tolerance of -0.005 mm, and includes 250 individual pieces.
  • Covers a diameter range from 5.00 mm to 9.98 mm.
  • Features a class ZZ tolerance of 0.0050 mm.
  • Includes 250 individual steel gages.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for NO-GO gaging inspection.
  • Ideal for gaging slot widths and locations, and measuring hole diameters.
  • Comes with a certificate of accuracy with NIST traceable standards.
  • Manufactured in the United States.