Learn More
MSC Vermont Gage Plug Gage Set, Class ZZ, 0.0115-0.0605 in, 50 Pc
Catalog No. 5000529381
Encompass
The Vermont Gage Class ZZ Plug Gage Set is designed for precise measurements and inspections. This 50-piece set covers a diameter range from 0.0115 to 0.0605 inches with a tolerance of 0.000200 inches. Made of steel, these gages are suitable for various laboratory and industrial applications, and come with a Certificate of Accuracy with NIST Traceable Standards.
  • Includes 50 pieces for comprehensive gaging.
  • Covers a diameter range from 0.0115 to 0.0605 inches.
  • Maintains a precise tolerance of 0.000200 inches (Class ZZ).
  • Constructed from durable steel for longevity.
  • Supplied with a Certificate of Accuracy traceable to NIST standards.
  • Ideal for setting micrometers, gaging slot widths, and measuring hole diameters.
  • Supports GO gaging inspection and calculating distances between holes.
Catalog No. 50-005-29381 Supplier MSC Supplier No. 06463293
The Vermont Gage Class ZZ Plug Gage Set is designed for precise measurements and inspections. This 50-piece set covers a diameter range from 0.0115 to 0.0605 inches with a tolerance of 0.000200 inches. Made of steel, these gages are suitable for various laboratory and industrial applications, and come with a Certificate of Accuracy with NIST Traceable Standards.
  • Includes 50 pieces for comprehensive gaging.
  • Covers a diameter range from 0.0115 to 0.0605 inches.
  • Maintains a precise tolerance of 0.000200 inches (Class ZZ).
  • Constructed from durable steel for longevity.
  • Supplied with a Certificate of Accuracy traceable to NIST standards.
  • Ideal for setting micrometers, gaging slot widths, and measuring hole diameters.
  • Supports GO gaging inspection and calculating distances between holes.