Learn More
MSC Vermont Gage Plug Gage Set, Class ZZ, 0.061-0.75 in Range, 690 Pc
Catalog No. 5000533255
Encompass
This plug gage set is designed for precise measurements in various industrial and laboratory applications. It features a Class ZZ tolerance and covers a diameter range from 0.061 to 0.75 inch, providing 690 pieces for comprehensive gaging needs. Constructed from durable steel with a black oxide finish, this set ensures reliability and accuracy for critical measuring tasks.
  • Covers a diameter range from 0.061 to 0.75 in.
  • Features a class ZZ tolerance.
  • Constructed from steel with a black oxide finish.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Ideal for gaging slot widths, locations, and measuring hole diameters.
  • Includes a certificate of accuracy with NIST traceable standards.
  • Contains 690 pieces.
Catalog No. 50-005-33255 Supplier MSC Supplier No. 00413427
This plug gage set is designed for precise measurements in various industrial and laboratory applications. It features a Class ZZ tolerance and covers a diameter range from 0.061 to 0.75 inch, providing 690 pieces for comprehensive gaging needs. Constructed from durable steel with a black oxide finish, this set ensures reliability and accuracy for critical measuring tasks.
  • Covers a diameter range from 0.061 to 0.75 in.
  • Features a class ZZ tolerance.
  • Constructed from steel with a black oxide finish.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Ideal for gaging slot widths, locations, and measuring hole diameters.
  • Includes a certificate of accuracy with NIST traceable standards.
  • Contains 690 pieces.