Learn More
MSC Vermont Gage Plug Gage Set, Class ZZ, 0.251 to 0.5 in, 250 Pc
Catalog No. 5000513815
Encompass
This Class ZZ plug gage set provides a comprehensive solution for precision measurement. It includes 250 steel pieces with a range of 0.251 to 0.5 in and a tolerance of 0.000200 in. It is ideal for various applications such as setting micrometers, gaging slot widths, and measuring hole diameters, and comes with a certificate of accuracy.
  • Range of 0.251 to 0.5 in
  • Includes 250 pieces
  • Tolerance of 0.000200 in
  • Class ZZ
  • Plus tolerance type
  • Made of steel
  • Certificate of accuracy with NIST traceable standards included
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Suitable for gaging slot widths and locations
  • Suitable for measuring hole diameters
  • Suitable for GO gaging inspection
  • Suitable for calculating distances between holes
Catalog No. 50-005-13815 Supplier MSC Supplier No. 06463046
This Class ZZ plug gage set provides a comprehensive solution for precision measurement. It includes 250 steel pieces with a range of 0.251 to 0.5 in and a tolerance of 0.000200 in. It is ideal for various applications such as setting micrometers, gaging slot widths, and measuring hole diameters, and comes with a certificate of accuracy.
  • Range of 0.251 to 0.5 in
  • Includes 250 pieces
  • Tolerance of 0.000200 in
  • Class ZZ
  • Plus tolerance type
  • Made of steel
  • Certificate of accuracy with NIST traceable standards included
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Suitable for gaging slot widths and locations
  • Suitable for measuring hole diameters
  • Suitable for GO gaging inspection
  • Suitable for calculating distances between holes