Learn More
MSC Vermont Gage Plug Gage Set, Class ZZ, 0.626-0.75 in., 125 Piece
Catalog No. 5000529273
Encompass
This Class ZZ Plug Gage Set provides 125 pieces designed for precise measurement. With a range of 0.626 to 0.75 inches, this durable steel set ensures accurate performance for setting micrometers, gaging slot widths, and checking hole diameters.
  • Includes 125 individual gages for versatile measurement applications.
  • Enables quick and accurate measurements of hole sizes, slot widths, and distances between holes.
  • Supports setting micrometers, verniers, and Go gaging inspection.
  • Features a Class ZZ, Go (Plus) tolerance of 0.0002 "
  • Constructed from durable steel for enhanced wear life.
  • Comes with a certificate of accuracy and NIST traceable standards.
  • Gages are 2 inches long with a 10 microinch finish, round within 0.0001 "
Catalog No. 50-005-29273 Supplier MSC Supplier No. 06463087
This Class ZZ Plug Gage Set provides 125 pieces designed for precise measurement. With a range of 0.626 to 0.75 inches, this durable steel set ensures accurate performance for setting micrometers, gaging slot widths, and checking hole diameters.
  • Includes 125 individual gages for versatile measurement applications.
  • Enables quick and accurate measurements of hole sizes, slot widths, and distances between holes.
  • Supports setting micrometers, verniers, and Go gaging inspection.
  • Features a Class ZZ, Go (Plus) tolerance of 0.0002 "
  • Constructed from durable steel for enhanced wear life.
  • Comes with a certificate of accuracy and NIST traceable standards.
  • Gages are 2 inches long with a 10 microinch finish, round within 0.0001 "