Learn More
MSC Vermont Gage Plug & Pin Gage, 0.0345 in, Class ZZ Minus, 2 in
Catalog No. 5000528193
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 0.0345 inch diameter and an overall length of 2 inches. It is constructed from steel with a black oxide coating. This precision gage is designed for applications such as setting micrometers and for Go/No Go inspection, as well as use with verniers and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability.
  • Made of steel.
  • Features black oxide coating.
  • Suitable for setting micrometers.
  • Ideal for Go/No Go inspection.
  • Compatible with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Has a tolerance of 0.000200 "
Catalog No. 50-005-28193 Supplier MSC Supplier No. 50754928
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 0.0345 inch diameter and an overall length of 2 inches. It is constructed from steel with a black oxide coating. This precision gage is designed for applications such as setting micrometers and for Go/No Go inspection, as well as use with verniers and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability.
  • Made of steel.
  • Features black oxide coating.
  • Suitable for setting micrometers.
  • Ideal for Go/No Go inspection.
  • Compatible with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Has a tolerance of 0.000200 "