Learn More
MSC Vermont Gage Plug & Pin Gage, 0.2005 in, Class ZZ Minus, 10 Microfinish
Catalog No. 5000531108
Encompass
Vermont Gage Plug & Pin Gages are designed for precise measurement and inspection. This 0.2005 inch diameter gage is Class ZZ Minus and features a 10 microfinish, making it suitable for NoGo inspection in various applications.
  • Made from steel
  • Features a 10 microfinish
  • Overall length is 2 inches
  • Ideal for NoGo inspection
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-31108 Supplier MSC Supplier No. 89020051
Vermont Gage Plug & Pin Gages are designed for precise measurement and inspection. This 0.2005 inch diameter gage is Class ZZ Minus and features a 10 microfinish, making it suitable for NoGo inspection in various applications.
  • Made from steel
  • Features a 10 microfinish
  • Overall length is 2 inches
  • Ideal for NoGo inspection
  • Includes certificate of accuracy with NIST traceability