Learn More
MSC Vermont Gage Plug & Pin Gage, 0.2940 in, 2 in, ZZ Minus
Catalog No. 5000529754
Encompass
This precision Class ZZ Minus Plug & Pin Gage is engineered for accurate measurement tasks. It features a 0.2940 inch diameter and 2 inch overall length, making it suitable for applications such as setting micrometers, performing NoGo inspections, and calibrating various measuring devices. Constructed from durable steel, this gage is finished with black oxide for enhanced longevity.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Black oxide coating for enhanced durability.
  • Constructed from steel for robust performance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Diameter of 0.2940 inch ensures precise measurements.
  • Overall length of 2 inch for versatile use.
  • Tolerance of 0.000200 inch for high accuracy.
  • Class ZZ Minus for specific gauging requirements.
Catalog No. 50-005-29754 Supplier MSC Supplier No. 00530360
This precision Class ZZ Minus Plug & Pin Gage is engineered for accurate measurement tasks. It features a 0.2940 inch diameter and 2 inch overall length, making it suitable for applications such as setting micrometers, performing NoGo inspections, and calibrating various measuring devices. Constructed from durable steel, this gage is finished with black oxide for enhanced longevity.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Black oxide coating for enhanced durability.
  • Constructed from steel for robust performance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Diameter of 0.2940 inch ensures precise measurements.
  • Overall length of 2 inch for versatile use.
  • Tolerance of 0.000200 inch for high accuracy.
  • Class ZZ Minus for specific gauging requirements.