Learn More
MSC Vermont Gage Plug & Pin Gage, 0.5180 in Diameter, Class ZZ Minus
Catalog No. 5000531467
Encompass
This Class ZZ Minus Plug & Pin Gage from Vermont Gage features a 0.5180 in diameter and a 2 in overall length, constructed from durable steel. It is designed for precise measurement applications such as setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability, ensuring reliable performance.
  • Precise 0.5180 in diameter.
  • 2 in overall length.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a 0.000200 in tolerance.
Catalog No. 50-005-31467 Supplier MSC Supplier No. 89051809
This Class ZZ Minus Plug & Pin Gage from Vermont Gage features a 0.5180 in diameter and a 2 in overall length, constructed from durable steel. It is designed for precise measurement applications such as setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability, ensuring reliable performance.
  • Precise 0.5180 in diameter.
  • 2 in overall length.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a 0.000200 in tolerance.