Learn More
MSC Vermont Gage Plug & Pin Gage, 11.32 mm, Class ZZ Minus
Catalog No. 5000529112
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision tool designed for accurate measurement and inspection. It is ideal for verifying dimensional tolerances in manufacturing, machining, and engineering applications.
  • Designed for precise measurement and inspection
  • Class ZZ Minus tolerance for high accuracy
  • 11.32 mm diameter for measuring holes, distances, and slots
  • 2 inch overall length
  • 10 microfinish
Catalog No. 50-005-29112 Supplier MSC Supplier No. 40438772
This Class ZZ Minus Plug & Pin Gage is a precision tool designed for accurate measurement and inspection. It is ideal for verifying dimensional tolerances in manufacturing, machining, and engineering applications.
  • Designed for precise measurement and inspection
  • Class ZZ Minus tolerance for high accuracy
  • 11.32 mm diameter for measuring holes, distances, and slots
  • 2 inch overall length
  • 10 microfinish