Learn More
MSC Vermont Gage Plug & Pin Gage, 14.94 mm, Class ZZ Minus, Steel
Catalog No. 5000527041
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise Go/No-Go inspection. It features a 14.94 mm diameter with a 0.00508 mm tolerance and a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Made from steel for durability
  • Suitable for NoGo inspection applications
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a 10 micro finish
  • Measures 2 inches in overall length
  • Provides a tolerance of 0.00508 mm
  • Designed with a 14.94 mm diameter measurement
Catalog No. 50-005-27041 Supplier MSC Supplier No. 40442394
This Class ZZ Minus Plug & Pin Gage is designed for precise Go/No-Go inspection. It features a 14.94 mm diameter with a 0.00508 mm tolerance and a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Made from steel for durability
  • Suitable for NoGo inspection applications
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a 10 micro finish
  • Measures 2 inches in overall length
  • Provides a tolerance of 0.00508 mm
  • Designed with a 14.94 mm diameter measurement