Learn More
MSC Vermont Gage Plug & Pin Gage, 2.06mm, Class ZZ Minus, 10 Micro
Catalog No. 5000527851
Encompass
This is a Vermont Gage Class ZZ Minus Plug & Pin Gage, specifically designed for accurate measurement and inspection. It features a 2.06 mm diameter, a 2 in overall length, and a 10 microfinish, making it suitable for a variety of precision applications.
  • Class ZZ Minus tolerance
  • 2.06 mm diameter
  • 2 in overall length
  • 10 microfinish for smooth surface
  • Suitable for precise measurement and inspection
Catalog No. 50-005-27851 Supplier MSC Supplier No. 40429516
This is a Vermont Gage Class ZZ Minus Plug & Pin Gage, specifically designed for accurate measurement and inspection. It features a 2.06 mm diameter, a 2 in overall length, and a 10 microfinish, making it suitable for a variety of precision applications.
  • Class ZZ Minus tolerance
  • 2.06 mm diameter
  • 2 in overall length
  • 10 microfinish for smooth surface
  • Suitable for precise measurement and inspection