Learn More
MSC Vermont Gage Plug & Pin Gage, 20.99 mm Diameter, Class ZZ Minus, 2 Inch OAL
Catalog No. 5000527148
Encompass
This precision plug and pin gage is designed for accurate measurement and inspection. Featuring a 20.99 mm diameter and a 2-inch overall length, this Class ZZ Minus gage is made from durable steel and includes a certificate of accuracy with NIST traceability. It is ideal for setting micrometers, performing NoGo inspections, and use with other measuring devices.
  • Precision Class ZZ Minus tolerance ensures high accuracy.
  • Constructed from durable steel for long-lasting performance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Perfect for NoGo inspection applications.
  • Compatible with various measuring devices.
Catalog No. 50-005-27148 Supplier MSC Supplier No. 40448441
This precision plug and pin gage is designed for accurate measurement and inspection. Featuring a 20.99 mm diameter and a 2-inch overall length, this Class ZZ Minus gage is made from durable steel and includes a certificate of accuracy with NIST traceability. It is ideal for setting micrometers, performing NoGo inspections, and use with other measuring devices.
  • Precision Class ZZ Minus tolerance ensures high accuracy.
  • Constructed from durable steel for long-lasting performance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Perfect for NoGo inspection applications.
  • Compatible with various measuring devices.