Learn More
MSC Vermont Gage Plug & Pin Gage, .6390 inch, Class ZZ Minus, Tool Steel
Catalog No. 5000531087
Encompass
This standard Class ZZ Minus Plug & Pin Gage features a .6390 inch diameter, meticulously crafted from hardened tool steel. Designed for highly accurate measurements, it provides a Class ZZ tolerance and is NIST traceable, ensuring reliable performance in precision applications.
  • Manufactured from hardened tool steel (60/65 Rc)
  • Class ZZ tolerance of .0002 inch or .005 mm
  • Two inch overall length
  • Surface finish of 10 microinch or better
  • Roundness within .0001 inch or .0025 mm
  • Marked with size and direction of tolerance (for gages over .0605 inch)
  • NIST traceable
  • Includes a certificate of accuracy
Catalog No. 50-005-31087 Supplier MSC Supplier No. 89063903
This standard Class ZZ Minus Plug & Pin Gage features a .6390 inch diameter, meticulously crafted from hardened tool steel. Designed for highly accurate measurements, it provides a Class ZZ tolerance and is NIST traceable, ensuring reliable performance in precision applications.
  • Manufactured from hardened tool steel (60/65 Rc)
  • Class ZZ tolerance of .0002 inch or .005 mm
  • Two inch overall length
  • Surface finish of 10 microinch or better
  • Roundness within .0001 inch or .0025 mm
  • Marked with size and direction of tolerance (for gages over .0605 inch)
  • NIST traceable
  • Includes a certificate of accuracy