Learn More
MSC Vermont Gage Plug & Pin Gage, 8.88 mm, Steel, Class ZZ Minus
Catalog No. 5000532708
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements, ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. It features an 8.88 mm diameter, 2 in overall length, and is constructed from durable steel with a 10 Micro finish, offering a tolerance of 0.00508 mm.
  • Designed for quick and accurate measurements
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Diameter of 8.88 mm
  • Overall length of 2 in
  • Constructed from steel
  • Features a 10 Micro finish
  • Tolerance of 0.00508 mm
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-32708 Supplier MSC Supplier No. 40436339
This Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements, ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. It features an 8.88 mm diameter, 2 in overall length, and is constructed from durable steel with a 10 Micro finish, offering a tolerance of 0.00508 mm.
  • Designed for quick and accurate measurements
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Diameter of 8.88 mm
  • Overall length of 2 in
  • Constructed from steel
  • Features a 10 Micro finish
  • Tolerance of 0.00508 mm
  • Includes certificate of accuracy with NIST traceability