Learn More
MSC Vermont Gage Plug & Pin Gage, 9.24 mm, ZZ Minus, 0.00508 mm Tolerance
Catalog No. 5000528456
Encompass
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 9.24 mm diameter with a Class ZZ Minus rating and a tight tolerance of 0.00508 mm, making it suitable for demanding applications in quality control and calibration.
  • Measures 9.24 mm
  • Overall length of 2 in
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Suitable for NoGo inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microfinish
  • Class ZZ Minus
Catalog No. 50-005-28456 Supplier MSC Supplier No. 40436693
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 9.24 mm diameter with a Class ZZ Minus rating and a tight tolerance of 0.00508 mm, making it suitable for demanding applications in quality control and calibration.
  • Measures 9.24 mm
  • Overall length of 2 in
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Suitable for NoGo inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microfinish
  • Class ZZ Minus