Learn More
MSC Vermont Gage Plug & Pin Gage, Black Guard, Class ZZ Minus, 8.77 mm
Catalog No. 5000533982
Encompass
This Class ZZ Minus Plug & Pin Gage features a black oxide finish for enhanced corrosion prevention and visual wear indication. As the gage wears, its black surface turns silver, signaling potential calibration or replacement needs. Designed for precision measurement, it is suitable for NoGo inspection and setting various measuring devices. It has an 8.77 mm diameter and an overall length of 2 in, made from durable steel.
  • Provides enhanced corrosion prevention and visual wear indication.
  • Black oxide finish reduces friction and extends gage life.
  • Suitable for NoGo inspection and setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 microfinish and is constructed from steel.
  • Class ZZ Minus tolerance ensures precision.
Catalog No. 50-005-33982 Supplier MSC Supplier No. 40436222
This Class ZZ Minus Plug & Pin Gage features a black oxide finish for enhanced corrosion prevention and visual wear indication. As the gage wears, its black surface turns silver, signaling potential calibration or replacement needs. Designed for precision measurement, it is suitable for NoGo inspection and setting various measuring devices. It has an 8.77 mm diameter and an overall length of 2 in, made from durable steel.
  • Provides enhanced corrosion prevention and visual wear indication.
  • Black oxide finish reduces friction and extends gage life.
  • Suitable for NoGo inspection and setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 microfinish and is constructed from steel.
  • Class ZZ Minus tolerance ensures precision.