Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0050 in, 2 in
Catalog No. 5000531947
Encompass
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.0050 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Ships with a rust preventative coating in a sealed plastic bag.
Catalog No. 50-005-31947 Supplier MSC Supplier No. 94360583
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.0050 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Ships with a rust preventative coating in a sealed plastic bag.