Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0175 in, 2 in OAL
Catalog No. 5000531739
Encompass
This precision measuring tool is designed for accurate inspection, featuring a 0.0175 inch diameter and 2 inch overall length. Manufactured from durable steel with a black oxide finish, it is ideal for setting micrometers, NoGo inspection, and use with other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability for reliable measurements.
  • High accuracy with class ZZ Minus tolerance.
  • Durable steel construction with black oxide finish.
  • Certified accuracy with NIST traceability.
  • Versatile for setting micrometers, NoGo inspection, and other measuring devices.
  • Optimal dimensions: 0.0175 inch diameter and 2 inch overall length.
  • Proudly made in the USA.
Catalog No. 50-005-31739 Supplier MSC Supplier No. 62262753
This precision measuring tool is designed for accurate inspection, featuring a 0.0175 inch diameter and 2 inch overall length. Manufactured from durable steel with a black oxide finish, it is ideal for setting micrometers, NoGo inspection, and use with other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability for reliable measurements.
  • High accuracy with class ZZ Minus tolerance.
  • Durable steel construction with black oxide finish.
  • Certified accuracy with NIST traceability.
  • Versatile for setting micrometers, NoGo inspection, and other measuring devices.
  • Optimal dimensions: 0.0175 inch diameter and 2 inch overall length.
  • Proudly made in the USA.