Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0185 in Diameter, 2 in Length, Steel
Catalog No. 5000531718
Encompass
Vermont Gage Class ZZ Minus Plug & Pin Gages are precision tools made from steel, designed for accurate measurement applications. They are suitable for setting micrometers, NoGo inspections, and use with verniers and other measuring devices, ensuring high accuracy.
  • Precision ground steel construction
  • Suitable for setting micrometers, for NoGo inspection, and for use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Provides a tolerance of 0.000200 inch
Catalog No. 50-005-31718 Supplier MSC Supplier No. 89001853
Vermont Gage Class ZZ Minus Plug & Pin Gages are precision tools made from steel, designed for accurate measurement applications. They are suitable for setting micrometers, NoGo inspections, and use with verniers and other measuring devices, ensuring high accuracy.
  • Precision ground steel construction
  • Suitable for setting micrometers, for NoGo inspection, and for use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Provides a tolerance of 0.000200 inch