Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0265 in Diameter, 2 in Overall Length
Catalog No. 5000513986
Encompass
This plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, verniers, and other measuring devices. It features a 0.0265 inch diameter and a 2 inch overall length, constructed from steel with a 10 micro coating/finish.
  • Includes certificate of accuracy with NIST traceability.
  • Made from steel.
  • Suitable for setting micrometers.
  • Suitable for NoGo inspection.
  • Suitable for verniers and other measuring devices.
  • Features a 10 micro coating/finish.
  • Country of origin: United States.
Catalog No. 50-005-13986 Supplier MSC Supplier No. 89002653
This plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, verniers, and other measuring devices. It features a 0.0265 inch diameter and a 2 inch overall length, constructed from steel with a 10 micro coating/finish.
  • Includes certificate of accuracy with NIST traceability.
  • Made from steel.
  • Suitable for setting micrometers.
  • Suitable for NoGo inspection.
  • Suitable for verniers and other measuring devices.
  • Features a 10 micro coating/finish.
  • Country of origin: United States.