Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0300 in, 2 in Long
Catalog No. 5000515417
Encompass
This Class ZZ Minus plug and pin gage is engineered for precise measurement and calibration tasks, including setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. It is fabricated from 52100 carbon chrome hardened steel and features a 0.0300 in diameter with a 2 in overall length, designed to a tolerance of -0.0002 to +0.0000 in.
  • Accurate for setting micrometers and calibrating various measuring instruments.
  • Manufactured from durable 52100 carbon chrome hardened steel, hardened to 60/62rc.
  • Features a 0.0300 in diameter and a 2 in overall length.
  • Tolerance of -0.0002 to +0.0000 in for precision applications.
  • Includes a certificate of accuracy with NIST traceability.
  • 10 micro coating/finish ensures consistent performance.
Catalog No. 50-005-15417 Supplier MSC Supplier No. 89003008
This Class ZZ Minus plug and pin gage is engineered for precise measurement and calibration tasks, including setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. It is fabricated from 52100 carbon chrome hardened steel and features a 0.0300 in diameter with a 2 in overall length, designed to a tolerance of -0.0002 to +0.0000 in.
  • Accurate for setting micrometers and calibrating various measuring instruments.
  • Manufactured from durable 52100 carbon chrome hardened steel, hardened to 60/62rc.
  • Features a 0.0300 in diameter and a 2 in overall length.
  • Tolerance of -0.0002 to +0.0000 in for precision applications.
  • Includes a certificate of accuracy with NIST traceability.
  • 10 micro coating/finish ensures consistent performance.