Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0450 in Diameter, 2 in OAL
Catalog No. 5000531846
Encompass
This Class ZZ Minus Plug & Pin Gage is a high-precision tool. Constructed from steel, it offers robust durability for various measurement tasks. This versatile gage is suitable for a range of metrology applications, including setting micrometers, conducting NoGo inspections, and use with verniers and other measuring devices.
  • High-precision tool with Class ZZ Minus designation
  • Constructed from steel for robust durability
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish for precision and extended operational life
  • Suitable for setting micrometers
  • Suitable for NoGo inspection
  • Suitable for use with verniers and other measuring devices
Catalog No. 50-005-31846 Supplier MSC Supplier No. 89004501
This Class ZZ Minus Plug & Pin Gage is a high-precision tool. Constructed from steel, it offers robust durability for various measurement tasks. This versatile gage is suitable for a range of metrology applications, including setting micrometers, conducting NoGo inspections, and use with verniers and other measuring devices.
  • High-precision tool with Class ZZ Minus designation
  • Constructed from steel for robust durability
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish for precision and extended operational life
  • Suitable for setting micrometers
  • Suitable for NoGo inspection
  • Suitable for use with verniers and other measuring devices