Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0470 in, 2 in, Steel
Catalog No. 5000531693
Encompass
This Class ZZ Minus plug and pin gage features a diameter of 0.0470 inches and an overall length of 2 inches. It is constructed from steel and has a tolerance of 0.000200 inches, with a 10 Micro coating/finish. This gage is designed for precise measurements and calibration tasks.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-31693 Supplier MSC Supplier No. 89004709
This Class ZZ Minus plug and pin gage features a diameter of 0.0470 inches and an overall length of 2 inches. It is constructed from steel and has a tolerance of 0.000200 inches, with a 10 Micro coating/finish. This gage is designed for precise measurements and calibration tasks.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.