Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0510 in Diameter, 2 in OAL
Catalog No. 5000531836
Encompass
This plug and pin gage is designed for precision measurement applications, featuring a 0.0510 inch diameter and a 2 inch overall length. Manufactured from steel with a black oxide coating, it provides reliable performance for various inspection tasks.
  • Precisely measures 0.0510 inch diameter
  • Overall length of 2 inches
  • Maintains a tight tolerance of 0.000200 inch
  • Constructed from durable steel
  • Features a protective black oxide coating
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Comes with a certificate of accuracy including NIST traceability
Catalog No. 50-005-31836 Supplier MSC Supplier No. 59893735
This plug and pin gage is designed for precision measurement applications, featuring a 0.0510 inch diameter and a 2 inch overall length. Manufactured from steel with a black oxide coating, it provides reliable performance for various inspection tasks.
  • Precisely measures 0.0510 inch diameter
  • Overall length of 2 inches
  • Maintains a tight tolerance of 0.000200 inch
  • Constructed from durable steel
  • Features a protective black oxide coating
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Comes with a certificate of accuracy including NIST traceability