Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0530 in Dia, 2 in OAL
Catalog No. 5000532145
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. It features a 0.0530 inch diameter and two inch overall length, made from steel, and has a 10 micro finish. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro finish.
  • Constructed from steel.
  • Provides a tolerance of 0.000200 "
Catalog No. 50-005-32145 Supplier MSC Supplier No. 89005300
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. It features a 0.0530 inch diameter and two inch overall length, made from steel, and has a 10 micro finish. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro finish.
  • Constructed from steel.
  • Provides a tolerance of 0.000200 "