Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0645 in Diameter, 2 in OAL
Catalog No. 5000535790
Encompass
The Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 10 Micro coating/finish and includes a Certificate of Accuracy with NIST Traceability. Made of steel, this gage is suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • 10 Micro coating/finish.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from steel.
  • Class ZZ Minus.
  • 0.0645 inch diameter.
  • 2 inch overall length.
  • 0.000200 decimal inch tolerance.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
Catalog No. 50-005-35790 Supplier MSC Supplier No. 89006456
The Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 10 Micro coating/finish and includes a Certificate of Accuracy with NIST Traceability. Made of steel, this gage is suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • 10 Micro coating/finish.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from steel.
  • Class ZZ Minus.
  • 0.0645 inch diameter.
  • 2 inch overall length.
  • 0.000200 decimal inch tolerance.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.