Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0820 in Dia, 2 in OAL
Catalog No. 5000529263
Encompass
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.0820 inch diameter, 2 inch overall length, and is constructed from steel with a black oxide finish. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Class ZZ Minus for precise measurements.
  • Diameter of 0.0820 "
  • Overall length of 2 "
  • Made from steel.
  • Black oxide finish for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Tolerance of 0.000200 "
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Compatible with verniers and other measuring devices.
Catalog No. 50-005-29263 Supplier MSC Supplier No. 06459374
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.0820 inch diameter, 2 inch overall length, and is constructed from steel with a black oxide finish. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Class ZZ Minus for precise measurements.
  • Diameter of 0.0820 "
  • Overall length of 2 "
  • Made from steel.
  • Black oxide finish for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Tolerance of 0.000200 "
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Compatible with verniers and other measuring devices.