Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0880 in Dia, 2 in OAL
Catalog No. 5000531138
Encompass
This plug and pin gage is designed for precise measurement applications. Made from durable steel and finished with black oxide, it offers reliability for various inspection tasks. It is ideal for calibrating measuring instruments and performing 'NoGo' inspections.
  • Black oxide coating.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Constructed from steel for durability.
Catalog No. 50-005-31138 Supplier MSC Supplier No. 88065727
This plug and pin gage is designed for precise measurement applications. Made from durable steel and finished with black oxide, it offers reliability for various inspection tasks. It is ideal for calibrating measuring instruments and performing 'NoGo' inspections.
  • Black oxide coating.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Constructed from steel for durability.