Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.0990 in Diameter, 2 in OAL
Catalog No. 5000530509
Encompass
The Vermont Gage Plug & Pin Gage is designed for precise measurement applications. This Class ZZ Minus gage has a 0.0990 in diameter and an overall length of 2 in, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. It is constructed from steel and comes with a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from steel.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-30509 Supplier MSC Supplier No. 89009906
The Vermont Gage Plug & Pin Gage is designed for precise measurement applications. This Class ZZ Minus gage has a 0.0990 in diameter and an overall length of 2 in, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. It is constructed from steel and comes with a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from steel.
  • Features a 10 micro coating/finish.