Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1070 in Diameter, 2 in Length
Catalog No. 5000515610
Encompass
This plug and pin gage is designed for precise measurement and inspection tasks, such as setting micrometers, conducting no-go inspections, and calibrating verniers and other measuring devices. Constructed from durable steel with a 10 micro coating/finish, it offers a diameter of 0.1070 inches and an overall length of 2 inches, ensuring high accuracy with a tolerance of 0.000200 inches. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Designed for setting micrometers, no-go inspection, and calibrating measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.
  • Measures 0.1070 in diameter and 2 in overall length.
  • Provides a tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15610 Supplier MSC Supplier No. 89010706
This plug and pin gage is designed for precise measurement and inspection tasks, such as setting micrometers, conducting no-go inspections, and calibrating verniers and other measuring devices. Constructed from durable steel with a 10 micro coating/finish, it offers a diameter of 0.1070 inches and an overall length of 2 inches, ensuring high accuracy with a tolerance of 0.000200 inches. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Designed for setting micrometers, no-go inspection, and calibrating measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.
  • Measures 0.1070 in diameter and 2 in overall length.
  • Provides a tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability.