Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1175 in Dia, 2 in OAL
Catalog No. 5000530224
Encompass
This plug and pin gage is designed for precise measurement and inspection applications. Crafted from durable steel with a 10 micro finish, it features a 0.1175 inch diameter and a 2 inch overall length, adhering to Class ZZ Minus tolerance standards. It is ideal for calibrating micrometers, conducting NoGo inspections, and verifying other measuring devices.
  • Designed for precise measurement and inspection applications
  • Crafted from durable steel
  • Features a 0.1175 inch diameter
  • Has a 2 inch overall length
  • Adheres to class ZZ Minus tolerance standards
  • Equipped with a 10 micro coating or finish
  • Ideal for calibrating micrometers
  • Suitable for no-go inspection
  • Useful for verifying verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-30224 Supplier MSC Supplier No. 89011753
This plug and pin gage is designed for precise measurement and inspection applications. Crafted from durable steel with a 10 micro finish, it features a 0.1175 inch diameter and a 2 inch overall length, adhering to Class ZZ Minus tolerance standards. It is ideal for calibrating micrometers, conducting NoGo inspections, and verifying other measuring devices.
  • Designed for precise measurement and inspection applications
  • Crafted from durable steel
  • Features a 0.1175 inch diameter
  • Has a 2 inch overall length
  • Adheres to class ZZ Minus tolerance standards
  • Equipped with a 10 micro coating or finish
  • Ideal for calibrating micrometers
  • Suitable for no-go inspection
  • Useful for verifying verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability