Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1220 in Dia, 2 in OAL, Steel
Catalog No. 5000531751
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is engineered for precise measurement applications, featuring a 0.1220 inch diameter and a 2 inch overall length. Constructed from durable steel, it is an essential tool for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection tasks.
  • Designed for precise measurement applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Effective for NoGo inspection
  • Constructed from durable steel
  • Features a 0.1220 in diameter
  • Has an overall length of 2 in
  • Includes a tolerance of 0.000200 in
  • Features a 10 micro coating/finish
  • Provided with a certificate of accuracy with NIST traceability
Catalog No. 50-005-31751 Supplier MSC Supplier No. 62257167
This Vermont Gage Class ZZ Minus Plug & Pin Gage is engineered for precise measurement applications, featuring a 0.1220 inch diameter and a 2 inch overall length. Constructed from durable steel, it is an essential tool for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection tasks.
  • Designed for precise measurement applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Effective for NoGo inspection
  • Constructed from durable steel
  • Features a 0.1220 in diameter
  • Has an overall length of 2 in
  • Includes a tolerance of 0.000200 in
  • Features a 10 micro coating/finish
  • Provided with a certificate of accuracy with NIST traceability